Learn More
ATAD2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546087
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PGAGAEPGAT GGSSHFISSR TRSSKTRAAS CPAAKAGGSG GAGVTLDEAR Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%
ATAD2B Polyclonal antibody specifically detects ATAD2B in Human samples. It is validated for Western Blot.Specifications
ATAD2B | |
Polyclonal | |
Unconjugated | |
ATAD2B | |
1110014E10Rik; ANCCA; ATAD2; Atad2b; ATPase family AAA domain containing 2B; ATPase family AAA domain-containing protein 2B; ATPase family, AAA domain containing 2B; BC032887; C79189; D530031C13Rik; KIAA1240; L16; PRO2000 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54454 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9ULI0 | |
ATAD2B | |
Synthetic peptide directed towards the N-terminal of human ATAD2B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |