missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAH2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547509
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse ASAH2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ASAH2 Polyclonal antibody specifically detects ASAH2 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
ASAH2 | |
Polyclonal | |
Unconjugated | |
Asah2 | |
acylsphingosine deacylase 2; AI585898; Asah2; BCDase; hCD; HNAC1; LCDase; mitochondrial ceramidase; N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2; N-acylsphingosine amidohydrolase 2; NCDase; N-CDase; Neutral ceramidase; Neutral ceramidase soluble form; neutral/alkaline ceramidase; non-lysosomal ceramidase | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
56624 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9NR71 | |
Asah2 | |
Mouse myeloma cell line NS0-derived recombinant human ASAH2/N-acylsphingosine Amidohydrolase-2 Thr15-Ile761 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |