Learn More
Arylsulfatase A/ARSA Mouse anti-Human, Clone: 324802, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2485
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439475
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 28-02-2020
Description
Description
Arylsulfatase A/ARSA Monoclonal antibody MAB2485 Specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Arylsulfatase A/ARSA
324802
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ARSA
Mouse myeloma cell line NS0-derived recombinant human Arylsulfatase A/ARSA Arg19-Ala507 Accession # AAH14210
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
410
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
AAH14210
ARSA, arylsulfatase A, ASA, Cerebroside 3-sulfatase, cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Arylsulfatase A/ARSA in direct ELISAs and Western blots. In direct ELISAs and Western blots, 20-50% cross-reactivity with recombinant mouse ARSA is observed.