missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547350
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Artemin is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Artemin Polyclonal antibody specifically detects Artemin in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Artemin | |
Polyclonal | |
Unconjugated | |
ARTN | |
Artemin; ARTN; Enovin; EVN; NBN; neublastin; neurotrophic factor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9048 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q5T4W7 | |
ARTN | |
E. coli-derived recombinant human Artemin Ala108-Gly220 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |