Learn More
Artemin Mouse anti-Human, Clone: 314908, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2589
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459555
  • £249.49 / 100µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Artemin Monoclonal antibody MAB2589 Specifically detects Artemin in Human samples. It is validated for Western Blotting
Specifications
Specifications
Artemin
314908
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ARTN
E. coli-derived recombinant human Artemin Ala108-Gly220 Accession # Q5T4W7.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9048
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q5T4W7.1
artemin, ARTN, ENOVIN, EVN, EVNneurotrophic factor, NBN, Neublastin
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Artemin in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BIG-H3, rhCripto-1, recombinant mouse (rm) Cripto-1, recombinant Drosophila DPP, rhDPP-6, rhGDNF, rhLAP, rhLatent TGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 3, rhLefty, rmLefty-1, rhMIS, rhNeurturin, rmNODAL, rhPersephin, recombinant amphibian TGF-beta 5, rhTGF-alpha or rhTGF-beta 2 is observed.