Learn More
Arylsulfatase B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527794
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, HepG2, ARSB tansfected 293T cell, mouse placenta, rat testis. Predicted reactivity: Dog (86%), Cat (89%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Arylsulfatase B Polyclonal antibody specifically detects Arylsulfatase B in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Arylsulfatase B | |
Polyclonal | |
Unconjugated | |
ARSB | |
1110007C02Rik; AI480648; Arsb; Arylsulfatase B; Arylsulfatase B (MPS VI); arylsulfatase B complex; arylsulfatase B regulation; arylsulfatase B structural; arylsulfatase B temporal regulation; As1; As-1; As-1r; As1-r; As-1s; As1-s; As-1t; As1-t; ASB; Asr-1; Ast-1; G4S; MPS6; N-acetylgalactosamine-4-sulfatase | |
Rabbit | |
Protein A | |
RUO | |
11881, 25227, 411 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
6.11 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P15848, P50429, P50430 | |
ARSB | |
Recombinant fragment corresponding to a region within amino acids 250 and 529 of Human ARSB | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |