Learn More
Arylsulfatase A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528372
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (89%), Rat (88%), Dog (91%), Pig (92%), Chimpanzee (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Arylsulfatase A Polyclonal antibody specifically detects Arylsulfatase A in Human samples. It is validated for Western Blot.Specifications
Arylsulfatase A | |
Polyclonal | |
Unconjugated | |
ARSA | |
ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.44 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P15289 | |
ARSA | |
Recombinant fragment corresponding to a region within amino acids 169 and 405 of Human ARSA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |