missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-arrestin Monoclonal Antibody (PDS-1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA116962
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: retina lysate.
S-arrestin Monoclonal antibody specifically detects S-arrestin in Bovine, Human, Porcine samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
S-arrestin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P08168, P10523, P79260 | |
SAG | |
Porcine retinal arrestin S-antigen. | |
100 μL | |
Primary | |
Bovine, Human, Porcine | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
PDS-1 | |
Unconjugated | |
SAG | |
48 kDa protein; A930001K18Rik; Arr1; arrestin; arrestin 1; arrestin1; arrestin-1; DKFZp686D1084; DKFZp686I1383; Irbp; photoreceptor protein/alleged cGMP phosphodiesterase activation modulator; put. S-antigen C-terminus; retinal S antigen; retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; rod photoreceptor arrestin; RP47; Sag; S-AG; SAGMR; SANTI; S-antigen; S-antigen visual arrestin; S-antigen, retina and pineal gland (arrestin); S-antigen; retina and pineal gland (arrestin); S-arrestin; S-arrestin short form; visual arrestin 1 | |
Mouse | |
Protein G | |
RUO | |
280922, 397151, 6295 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |