Learn More
Rho guanine nucleotide exchange factor (GEF) 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ARHGEF1.
Brand: Abnova H00009138-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. [provided by RefSeq
Sequence: CRPGPEGQLAATALRKVLSLKQLLFPAEEDNGAGPPRDGDGVPGGGPLSPARTQEIQENLLSLEETMKQLEELEEEFCRLRPLLSQLGGNSVPQPGCTSpecifications
Rho guanine nucleotide exchange factor (GEF) 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ARHGEF1. | |
ARHGEF1 | |
GEF1/LBCL2/LSC/P115-RHOGEF/SUB1.5 | |
Mouse | |
50 μL | |
Cell Cycle | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC034013 | |
ARHGEF1 | |
ARHGEF1 (AAH34013, 830 a.a. ∼ 927 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
9138 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |