missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D4-GDI Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519352
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology.
D4-GDI Polyclonal antibody specifically detects D4-GDI in Human samples. It is validated for Western Blot.Specifications
D4-GDI | |
Polyclonal | |
Unconjugated | |
ARHGDIB | |
ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
397 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P52566 | |
ARHGDIB | |
Synthetic peptide sequence (TEKAPEPHVE) corresponding to the N-terminus amino acids of ARHGDIB | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |