Learn More
ARHGAP15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531530
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (87%), Rat (82%), Rhesus Monkey (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ARHGAP15 Polyclonal antibody specifically detects ARHGAP15 in Chicken, Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ARHGAP15 | |
Polyclonal | |
Unconjugated | |
ARHGAP15 | |
5830480G12Rik; ArhGAP15; BM-024; BM-030; BM046; BM-046; hypothetical protein; polymorphic marker clone XE09; RCJMB04_1j23; Rho GTPase activating protein 15; rho GTPase-activating protein 15; Rho-type GTPase-activating protein 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
424303, 55843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q53QZ3, Q5ZMM3 | |
ARHGAP15 | |
Recombinant fragment corresponding to a region within amino acids 1 and 211 of Human ARHGAP15 | |
100 μL | |
Primary | |
Chicken, Human | |
Antibody | |
IgG |