Learn More
ZEB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528221
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, 293T, HepG2, MDA-MB-231, Neuro2A, NIH-3T3, C2C12, ZEB1 shRNA-transfected HeLa. Predicted reactivity: Mouse (91%), Rat (89%), Chicken (90%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZEB1 Polyclonal antibody specifically detects ZEB1 in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
ZEB1 | |
Polyclonal | |
Unconjugated | |
ZEB1 | |
[delta]EF1; 3110032K11Rik; AREB6; BZP; Delta EF1; delta-crystallin enhancer binding factor 1; deltaEF1; DELTA-EF1; FECD6; LOW QUALITY PROTEIN: zinc finger E-box-binding homeobox 1; MEB1; MGC133261; Negative regulator of IL2; Nil2; NIL2A; NIL-2A; NIL-2-A; NIL-2-A zinc finger protein; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; PPCD3; RP11-472N13.4; Tcf18; TCF8; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Tw; twirler; ZEB; Zeb1; Zfhep; Zfhx1a; Zfx1a; Zfx1ha; zinc finger E-box binding homeobox 1; zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1a; zinc finger homeodomain enhancer-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21417, 6935 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P37275, Q64318 | |
ZEB1 | |
Recombinant fragment corresponding to a region within amino acids 254 and 586 of Human AREB6 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
- ZEB1 Polyclonal Antibody
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methylisothiazol-3( 2H)-one and 2-methylisothiazol-3(2H)-one