Learn More
APPL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530501
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG, MCF-7. Predicted reactivity: Mouse (99%), Zebrafish (86%), Xenopus laevis (89%), Pig (98%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
APPL1 Polyclonal antibody specifically detects APPL1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
APPL1 | |
Polyclonal | |
Unconjugated | |
APPL1 | |
2900057D21Rik; 7330406P05Rik; Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; adaptor protein APPL like; adaptor protein containing pH domain, PTB domain and leucine zipper motif 1; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1; AI585782; AKT2 interactor; APPL; Appl1; AW209077; BB022931; C88264; DCC-interacting protein 13-alpha; DIP13; DIP13 alpha; DIP13A; DIP13alpha; dip13-alpha; Kiaa1428; MODY14; RGD1309388; signaling adaptor protein DIP13alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26060, 72993 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8K3H0, Q9UKG1 | |
APPL1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 205 of Human APPL1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |