missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518978
Code : LD
Additional Details : Weight : 0.01000kg
Description
Apolipoprotein L4 Polyclonal antibody specifically detects Apolipoprotein L4 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Apolipoprotein L4 | |
Polyclonal | |
Unconjugated | |
APOL4 | |
APOL4; apolipoprotein L, 4; Apolipoprotein L4; apolipoprotein L-IV; APOLIV; APOL-IV; OTTHUMP00000028937 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
80832 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9BPW4 | |
APOL4 | |
Synthetic peptide sequence (ENLNELTHIHQSLK) corresponding to the C-terminus amino acids of APOL4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |