Learn More
ApoA1 Monoclonal Antibody (532), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514732
Code : LE
Additional Details : Weight : 0.01000kg
Description
By sandwich ELISA, MA5-14732 can be used to detect Apo A-1 from serum samples. To increase sensitivity of sandwich ELISAs with MA5-14732, a biotinylated detection antibody followed by Streptavidin-HRP is recommended. By Western blot, MA5-14732 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MA5-14732 is also recommended for detecting endogenous Apo A-1 by Western blot. By Western blot, MA5-14732 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MA5-14732 is also recommended for detecting endogenous Apo A-1 by Western blot. Product MA514732 is a smaller package size of MIA1405 (formerly sold as a Seradyn product).
ApoA1 Monoclonal antibody specifically detects ApoA1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot.Specifications
ApoA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P02647 | |
APOA1 | |
Purified human plasma Apo A-1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
532 | |
Unconjugated | |
APOA1 | |
1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7 | |
Mouse | |
Protein A | |
RUO | |
335 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |