missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC3G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540735
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AKIFRGQVYS ELKYHPEMRF FHWFSKWRKL HRDQEYEVTW YISWSPCTKC Sequence homology: Human: 100%; Pig: 79%
APOBEC3G Polyclonal antibody specifically detects APOBEC3G in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
APOBEC3G | |
Polyclonal | |
Unconjugated | |
APOBEC3G | |
A3G; APOBEC3G; APOBEC-related cytidine deaminase; APOBEC-related protein; APOBEC-related protein 9; apolipoprotein B editing enzyme catalytic polypeptide-like 3G; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3G; apolipoprotein B mRNA editing enzyme catalytic subunit 3G; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G; apolipoprotein B mRNA-editing enzyme catalytic polypeptide 3G; ARCD; ARP9; ARP-9; ASGPR2; ASGP-R2; bK150C2.7; CEM15; CEM-15; CLEC4H2; Deoxycytidine deaminase; dJ494G10.1; DNA dC->dU editing enzyme; DNA dC->dU-editing enzyme APOBEC-3G; FLJ12740; HBXBP; HL-2; MDS019; OTTHUMP00000028911; phorbolin-like protein MDS019 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
60489 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HC16 | |
APOBEC3G | |
Synthetic peptide directed towards the N-terminal of human APOBEC3G | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |