missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC3F Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526184
Code : LD
Additional Details : Weight : 0.01000kg
Description
APOBEC3F Polyclonal antibody specifically detects APOBEC3F in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
APOBEC3F | |
Polyclonal | |
Unconjugated | |
APOBEC3F | |
A3F; APOBEC3F; apolipoprotein B editing enzyme catalytic polypeptide-like 3F; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3F; apolipoprotein B mRNA editing enzyme catalytic subunit 3F; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F; ARP8; BK150C2.4.MRNA; DNA dC->dU-editing enzyme APOBEC-3F; induced upon T-cell activation; KA6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
200316 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q8IUX4 | |
APOBEC3F | |
KLH conjugated synthetic peptide between 22-48 amino acids from the N-terminal region of human APOBEC3F | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |