missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AOX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513659
Code : LE
Additional Details : Weight : 0.01000kg
Description
AOX1 Polyclonal antibody specifically detects AOX1 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
AOX1 | |
Polyclonal | |
Unconjugated | |
AOX1 | |
AI196512; AI255253; Aldehyde oxidase; aldehyde oxidase (female form); aldehyde oxidase 1; aldehyde oxidase 2; Ao; AOH1; Aox1; Aox-1; Aox2; Aox-2; Azaheterocycle hydroxylase; azaheterocycle hydroxylase 1; Moro; Retinal oxidase; Ro | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
316 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
1.6 mg/mL | |
PBS with 0.09% sodium azide | |
Q06278 | |
AOX1 | |
KLH conjugated synthetic peptide between 698-726 amino acids from the central region of human AOX1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |