missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AOC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535855
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Bovine (80%).
AOC2 Polyclonal antibody specifically detects AOC2 in Human samples. It is validated for Immunocytochemistry.Specifications
AOC2 | |
Polyclonal | |
Unconjugated | |
AOC2 | |
Amine oxidase [copper-containing]; amine oxidase copper containing 2; amine oxidase, copper containing 2; amine oxidase, copper containing 2 (retina-specific); AOC2; DAO2; RAO; retina-specific amine oxidase; retina-specific copper amine oxidase; Semicarbazide-sensitive amine oxidase; SSAO | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.94 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75106 | |
AOC2 | |
Recombinant fragment corresponding to a region within amino acids 404 and 684 of AOC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |