missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519694
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse, rat, sheep, chicken, cow, dog and Xenopus laevis based on sequence homology.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
Annexin A2 | |
Polyclonal | |
ANXA2 | |
ANX2, ANX2L4, CAL1H, LIP2, LPC2, LPC2D, P36, PAP-IV, annexin II; annexin-2; calpactin I heavy chain; calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; lipocortin II; placental anticoagulant protein IV; protein I | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
302 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
Unconjugated | |
P07355 | |
ANXA2 | |
Synthetic peptide conjugated to KLH derived from within residues 200 - 300 of Human Annexin II. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |