Learn More
Annexin A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522266
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: H1299, NIH-3T3, HeLa. Predicted reactivity: Mouse (86%), Rat (89%), Dog (92%), Cat (93%), Pig (87%), Rabbit (92%), Chimpanzee (99%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Annexin A1 Polyclonal antibody specifically detects Annexin A1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Annexin A1 | |
Polyclonal | |
Unconjugated | |
ANXA1 | |
ANAX1; Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; ANX1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; Phospholipase A2 inhibitory protein; RP11-71A24.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16952, 301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P04083, P10107 | |
ANXA1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 228 of Annexin A1 (Uniprot ID#P04083) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |