Learn More
ANKHD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530427
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HepG2, HCT116, Jurkat, Raji, NCI-H929. Predicted reactivity: Mouse (95%), Rat (95%), Dog (96%), Pig (96%), Chicken (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ANKHD1 Polyclonal antibody specifically detects ANKHD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ANKHD1 | |
Polyclonal | |
Unconjugated | |
ANKHD1 | |
1110004O12Rik; 4933432B13Rik; 9130019P20Rik; A530027J04Rik; A630021B20Rik; AA571404; ANKHD1; ankyrin repeat and KH domain containing 1; ankyrin repeat and KH domain-containing protein 1; HIV-1 Vpr-binding ankyrin repeat protein; hMASK; KIAA1085; MASK; MASK1; mFLJ00246; mKIAA1085; Multiple ankyrin repeats single KH domain; multiple ankyrin repeats, single KH-domain homolog; PP2500; VBARP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54882 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IWZ3 | |
ANKHD1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 391 of Human ANKHD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |