Learn More
ANGPTL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528058
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HepG2 conditioned medium. Predicted reactivity: Mouse (80%), Rat (80%), Pig (85%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ANGPTL3 Polyclonal antibody specifically detects ANGPTL3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ANGPTL3 | |
Polyclonal | |
Unconjugated | |
ANGPTL3 | |
ANG-5; angiopoietin 5; angiopoietin like 3; Angiopoietin-5; angiopoietin-like 3; angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5; ANGPTL3; ANGPTL3(17-221); ANGPTL3(17-224); ANL3; FHBL2; hypl; hypolipidemia; UNQ153/PRO179 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27329 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.11 mg/mL | |
PBS with no preservative | |
Q9Y5C1 | |
ANGPTL3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 351 of Human ANGPTL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |