missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-like Protein 7/ANGPTL7 Mouse anti-Human, Clone: 121408, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB914

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461986

  • £390.72 / 500µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Angiopoietin-like Protein 7/ANGPTL7 Monoclonal antibody MAB914 Specifically detects Angiopoietin-like Protein 7/ANGPTL7 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Angiopoietin-like Protein 7/ANGPTL7
121408
Western Blot 1 ug/mL
O43827
angiopoietin-like 7, Angiopoietin-like factor, Angiopoietin-like protein 7, angiopoietin-related protein 7, ANGPTL7, AngX, CDT6, CDT6angiopoietin-like factor (CDT6), Cornea-derived transcript 6 protein, dJ647M16.1, RP4-647M16.2
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Angiopoietin-like Protein 7/ANGPTL7 in direct ELISAs and Western blots. In Western blots, less than 3% cross-reactivity with recombinant human Angiopoietin-1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ANGPTL7
E. coli-derived recombinant human Angiopoietin-like Protein 7/ANGPTL7 Lys28-Pro346 Accession # O43827
Protein A or G purified from ascites
RUO
Primary
10218
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only