Learn More
ANGPTL3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547127
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-1, rhAngiopoietin-2, recombinant mouse Angiopoietin-3, rhAngiopoietin-4, and rhAngiopoietin-like factor/CDT6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ANGPTL3 Polyclonal antibody specifically detects ANGPTL3 in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
ANGPTL3 | |
Polyclonal | |
Unconjugated | |
ANGPTL3 | |
ANG-5; angiopoietin 5; angiopoietin like 3; Angiopoietin-5; angiopoietin-like 3; angiopoietin-like protein 3; Angiopoietin-related protein 3; ANGPT5; ANGPTL3; ANGPTL3(17-221); ANGPTL3(17-224); ANL3; FHBL2; hypl; hypolipidemia; UNQ153/PRO179 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
30924 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9R182 | |
ANGPTL3 | |
Sf 21-derived recombinant mouse Angiopoietin-like Protein 3/ANGPTL3 Ser17-Thr455 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |