missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-3 Rat anti-Mouse, Clone: 113504, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB738

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13451756

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Angiopoietin-3 Monoclonal antibody MAB738 Specifically detects Angiopoietin-3 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Angiopoietin-3
113504
Western Blot 1 ug/mL
Purified
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Angiopoietin-3 in direct ELISA and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-1, rhAngiopoietin-2, rhAngiopoietin-like factor, and rhAngiopoietin-4 is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Angpt4
E. coli-derived recombinant mouse Angiopoietin-3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
51378
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only