Learn More
Angiopoietin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129640
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is specific for mouse Angiopoietin-2 (mAng-2). Partial cross-reactivity is observed with human Angiopoietin-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Angiopoietin 2 Polyclonal antibody specifically detects Angiopoietin 2 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
Angiopoietin 2 | |
Polyclonal | |
Unconjugated | |
ANGPT2 | |
AGPT2; Ang2; Ang-2; angiopoietin 2; angiopoietin-2; angiopoietin-2a; angiopoietin-2B; ANGPT2; endothelial growth factor; Tie2-ligand | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
80 mg/mL | |
whole serum diluted in 0.02M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2 | |
O15123, O35608 | |
ANGPT2 | |
Synthetic peptide corresponding to a region near the N-terminus of mouse angiopoietin-2, conjugated to KLH | |
RUO | |
11601, 285 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |