Learn More
beta Amyloid, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA118318
Code : LD
Additional Details : Weight : 0.01000kg
Product Code. 11591982
  • £229.40 / 100┬ÁL
Estimated Shipment
23-05-2018
Add to basket
Description
Description
PA1-18318 detects APP from rat samples.PA1-18318 has been successfully used in immunohistochemistry procedures.The PA1-18318 immunogen is a synthetic peptide corresponding to residues H(44) M N V Q N G K W E S D P S G T K T C(62) of mouse APP isoform A, conjugated to Blue Carrier Protein.Reconstitute with 100 ul of distilled water.
Specifications
Specifications
Amyloid beta A4
Polyclonal
whole serum with no preservative
P08592
APP, ABPP, Alzheimer disease amyloid protein, Cerebral vascular amyloid peptide, CVAP, Protease nexin-II, PN-II, APPI, PreA4, A4, AD1
Rabbit
IgG
Avoid Freeze/Thaw Cycles, store at -20°C
Primary
54226
Immunohistochemistry
Unconjugated
App
Serum
APP
Synthetic peptide corresponding to residues H(44) M N V Q N G K W E S D P S G T K T C(62) of mouse APP isoform A.
100μL
Antibody
Polyclonal
Rat
Videos