Learn More
beta Amyloid, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA118317
Code : LD
Additional Details : Weight : 0.01000kg
Product Code. 11581982
  • £229.40 / 100┬ÁL
Estimated Shipment
20-07-2018
Add to basket
Description
Description
PA1-18317 detects APP from rat samples.PA1-18317 has been successfully used in immunohistochemistry procedures.The PA1-18317 immunogen is a synthetic peptides corresponding to residuess C-E(145) T H L H W H T V A K E T(157), C-H(388) A H F Q K A K E R L E A K H R E R(405), and C-K(724) K K Q Y T S I H H G V V E(737) of human APP isoform A, conjugated to KLH.Reconstitute with 100 ul of distilled water.
Specifications
Specifications
Amyloid beta A4
Polyclonal
whole serum with no preservative
P08592
APP, ABPP, Alzheimer disease amyloid protein, Cerebral vascular amyloid peptide, CVAP, Protease nexin-II, PN-II, APPI, PreA4, A4, AD1
Rabbit
IgG
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
54226
Immunohistochemistry
Unconjugated
App
Serum
APP
Synthetic peptides corresponding to residuess C-E(145) T H L H W H T V A K E T(157), C-H(388) A H F Q K A K E R L E A K H R E R(405), and C-K(724) K K
100μL
Antibody
Polyclonal
Rat
Videos