missing translation for 'onlineSavingsMsg'
Learn More

beta Amyloid, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSA00050G

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11530423

  • £257.00 / 500µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Reconstitute with 500 ul of distilled water.
Specifications

Specifications

Amyloid beta A4
Polyclonal
Unconjugated
APP
Purified
APP
A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein
Ammonium sulfate precipitation
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage
Primary
351
Western Blot
1mg/mL
Antibody with no preservative
P05067, P12023, P08592
Alzheimer disease amyloid A4 protein homolog, ABPP, APP, Amyloidogenic glycoprotein, AG, Amyloid beta A4 protein, amyloid beta (A4) precursor protein, AAA, AD1, PN2, ABPP, APPI, CVAP, ABETA, CTF gamma, Peptidase nexin-II, Alzheimer disease
Rabbit
IgG
500μg
Antibody
Polyclonal
Rat
Videos