missing translation for 'onlineSavingsMsg'
Learn More

beta Amyloid, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSA00049W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11520423

  • £257.00 / 100µL


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Reconstitute with 100 ul of distilled water.
Specifications

Specifications

Amyloid beta A4
Polyclonal
whole serum with no preservative
P05067, P12023, P08592
Alzheimer disease amyloid A4 protein homolog, ABPP, APP, Amyloidogenic glycoprotein, AG, Amyloid beta A4 protein, amyloid beta (A4) precursor protein, AAA, AD1, PN2, ABPP, APPI, CVAP, ABETA, CTF gamma, Peptidase nexin-II, Alzheimer disease
Rabbit
100μL
Antibody
Polyclonal
HumanþMurineþRat
ImmunohistochemistryþWestern Blot
Unconjugated
APP
Serum
APP
A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein was used as the antigen
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
351
Videos