Learn More
Aminopeptidase LRAP/ERAP2 Mouse anti-Human, Clone: 3F5, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3830
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430276
  • £311.64 / 100µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Aminopeptidase LRAP/ERAP2 Monoclonal antibody MAB3830 Specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Aminopeptidase LRAP/ERAP2
3F5
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ERAP2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Aminopeptidase LRAP/ERAP2 Accession # Q6P179
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
64167
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q6P179
EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, ERAP2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.-
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Aminopeptidase LRAP/ERAP2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human PILS is observed.