missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD249 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518393
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
CD249 Polyclonal antibody specifically detects CD249 in Human, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
CD249 | |
Polyclonal | |
Unconjugated | |
Enpep | |
6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2028, 64017 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P50123, Q07075 | |
Enpep | |
Synthetic peptide sequence (EQYQKTSLAQEKEK) corresponding to the internal amino acids of ENPEP | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |