Learn More
Aminoacylase/ACY1 Rat anti-Mouse, Clone: 475730, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2700
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469635
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Aminoacylase/ACY1 Monoclonal antibody MAB2700 Specifically detects Aminoacylase/ACY1 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Aminoacylase/ACY1
475730
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ACY1
Mouse myeloma cell line NS0-derived recombinant mouse Aminoacylase/ACY1 Met1-Ser408 Accession # NP_079647
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
95
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
NP_079647
ACY1, ACY-1, ACY1D, acylase, aminoacylase 1, aminoacylase-1, EC 3.5.1, EC 3.5.1.14, N-acyl-L-amino-acid amidohydrolase
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Aminoacylase/ACY1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human ACY1 is observed.