Learn More
alpha Tubulin Monoclonal Antibody (DM1A), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 62204
Code : PN
Additional Details : Weight : 0.01000kg
Description
62204 detects alpha tubulin in human, mouse, rat, bovine, porcine, chicken, amphibian, gerbil and guinea pig samples. 62204 has been successfully used in Western blot, immunohistochemistry, immunoprecipitation, electron microscopy, immunocytochemistry and immunofluorescent applications. The 62204 immunogen is native chick brain microtubules. This antibody recognizes an epitope within amino acids 426-450 of alpha tubulin.
alpha Tubulin Monoclonal antibody specifically detects alpha Tubulin in Amphibian, Bovine, Canine, Chicken, Gerbil, Guinea Pig, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot.Specifications
alpha Tubulin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P81948, P68366, P68368, Q5XIF6 | |
TUBA4A | |
Native chick brain microtubules | |
40 μL | |
Primary | |
Amphibian, Bovine, Canine, Chicken, Gerbil, Guinea Pig, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot | |
DM1A | |
Unconjugated | |
TUBA4A | |
&agr-tubulin 84B; 1t; a1t; ALPHA 84C; alpha tubulin; alpha tubulin 84B; alpha[[1]] tubulin; alpha[[1]]-tubulin; alpha1; alpha1 tubulin; alpha1t; alpha1tub; alpha1tub84B; alpha1-tubulin; alpha84B; alphaT; alphat-1; alphatub; alpha-tub; alphaTub1; alphatub84; alphaTUB84B; alpha-tub84B; alphaTub84B-PA; alphatubulin; alpha-tubulin; Alpha-tubulin 1; Alpha-tubulin 2; Alpha-tubulin 3; Alpha-tubulin 3C; alpha-tubulin 3C/D; Alpha-tubulin 3D; Alpha-tubulin 3E; alpha-tubulin 4; Alpha-tubulin 6; alphaTubulin 84B; alpha-Tubulin 84B; alpha-Tubulin at 84B; alpha-tubulin I; Alpha-tubulin II; alpha-tubulin III; Alpha-tubulin isotype M-alpha-1; alpha-tubulin isotype M-alpha-2; Alpha-tubulin isotype M-alpha-4; alpha-tubulin isotype M-alpha-6; alpha-tubulin TUB1; alpha-tubulin ubiquitous; alphaTubulin84B; Alpha-Tubulin84B; ALS22; anon-EST:Liang-1.59; anon-EST:Liang-2.30; AT25469p; aTub84B; a-tub84B; bA408E5.3; B-ALPHA-1; bcm948; BEST:LD32507; cb944; CG1913; CG1913-PA; chr3R:2914276..2914416; clone 1.59; c | |
Mouse | |
Protein G | |
RUO | |
100151951, 100734500, 100857247, 22145, 316531, 478918, 7277, 777775 | |
4° C | |
Liquid |