Learn More
alpha Synuclein Monoclonal Antibody (Syn 211), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 328100
Code : 50
Additional Details : Weight : 0.50000kg
Description
This antibody reacts with the alpha-synuclein protein. This antibody does not cross-react with beta- or gamma-synuclein or synoretin. During development reactivity was confirmed with human. Reactivity with other species has not been tested.
alpha Synuclein Monoclonal antibody specifically detects alpha Synuclein in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
alpha Synuclein | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P37840 | |
SNCA | |
Recombinant human alpha-synuclein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry, Western Blot | |
Syn 211 | |
Unconjugated | |
SNCA | |
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor) | |
Mouse | |
Affinity chromatography | |
RUO | |
6622 | |
-20°C | |
Liquid |