missing translation for 'onlineSavingsMsg'
Learn More

alpha Synuclein Monoclonal Antibody (4B12), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA190346

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11564813

  • £516.00 / 100µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Predicted molecular weight of 16 kDa.

alpha Synuclein Monoclonal antibody specifically detects alpha Synuclein in Human samples. It is validated for Dot Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

alpha Synuclein
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide
P37840
SNCA
Recombinant human alpha synuclein produced in E. coli.
100 μL
Primary
Human
Antibody
IgG1
Dot Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
4B12
Unconjugated
SNCA
Alpha synuclein; alphaSYN; alpha-synuclein; NACP; non A-beta component of AD amyloid; non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; PARK1; PARK4; PD1; Snca; Syn; synuclein alpha; synuclein alpha-140; synuclein, alpha; synuclein, alpha (non A4 component of amyloid precursor)
Mouse
Ammonium sulfate precipitation
RUO
6622
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers