missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin alpha-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517443
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
Catenin alpha-1 Polyclonal antibody specifically detects Catenin alpha-1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Catenin alpha-1 | |
Polyclonal | |
Unconjugated | |
CTNNA1 | |
[a]E-catenin; 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI988031; alpha E catenin; alpha E-catenin; alpha(E)-catenin; alpha-E-catenin; cadherin associated protein; Cadherin-associated protein; CAP102; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha; catenin (cadherin-associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin alpha 1; catenin alpha 1 S homeolog; catenin alpha-1; catenin, alpha 1; Catna1; cell-adhesion protein alphaE catenin; ctnna; ctnna1; ctnna1.S; MDPT2; Renal carcinoma antigen NY-REN-13; XELAEV_18020018mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12385, 1495, 307505 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
9 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P26231, P35221 | |
CTNNA1 | |
Synthetic peptide corresponding to the carboxy-terminal sequence of human a-E-Catenin | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |