missing translation for 'onlineSavingsMsg'
Learn More

CRYAB, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA116951

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11571812

  • £257.00 / 100µL


Estimated Shipment
22-06-2018
Add to basket

Description

Description

This antibody is expected to react with rat, ovine, rabbit, monkey, hamster, bovine and porcine based on sequence homology.
Specifications

Specifications

alpha B Crystallin
Polyclonal
Unconjugated
CRYAB
Purified
CRYAB
Synthetic peptide between amino acids 100-175 of the human Crystallin AB protein.
Antigen affinity chromatography
Store at 4° C, do not freeze
Primary
1410
ImmunocytochemistryþImmunofluorescenceþWestern Blot
0.9mg/mL
tris citrate with 0.1% sodium azide; pH 7-8
P02511
Alpha crystallin B chain antibody, CryAB antibody, Alpha (B)-crystallin antibody, Rosenthal fiber componet antibody, Heat-shock protein beta-5 antibod
Rabbit
IgG
100μL
Antibody
Polyclonal
HumanþMurine
Videos