Learn More
Alkaline Phosphatase/ALPL Rat anti-Human, Clone: 388816, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2909
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449775
  • £313.76 / 100µg
Estimated Shipment date
from Supplier 06-07-2020
Description
Description
Alkaline Phosphatase/ALPL Monoclonal antibody MAB2909 Specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Western Blotting
Specifications
Specifications
Alkaline Phosphatase/ALPL
388816
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ALPL
E. coli-derived recombinant human Alkaline Phosphatase/ALPL Leu18-Ser502 Accession # P05186
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
249
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P05186
Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Alkaline Phosphatase/ALPL in direct ELISAs and Western blots.