Learn More
ALK-1 Rat anti-Mouse, Clone: 143430, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB770
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13451846
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
ALK-1 Monoclonal antibody MAB770 Specifically detects ALK-1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
ALK-1
143430
Western Blot 1 ug/mL
Q61288
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse ALK-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Activin RIA, rhActivin RIB, or rhALK-1.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ACVRL1
Mouse myeloma cell line NS0-derived recombinant mouse ALK-1 Asp23-Pro119 Accession # Q61288
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
94
Reconstitute at 0.5 mg/mL in sterile PBS.