missing translation for 'onlineSavingsMsg'
Learn More

ALCAM/CD166 Rat anti-Mouse, Clone: 200622, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1172

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488405

  • £264.01 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

ALCAM/CD166 Monoclonal antibody MAB1172 Specifically detects ALCAM/CD166 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

ALCAM/CD166
200622
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ALCAM
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166, Trp28-Lys527, Accession # AAC06342
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
214
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
AAC06342
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse ALCAM/CD166 in Western blots and direct ELISAs. In Western blots, shows 100% cross-reactivity with recombinant human (rh) ALCAM and no cross-reactivity with rhBCAM, rhEpCAM, recombinant mouse (rm) MAdCAM-1, rhMCAM, rmNCAM-L1, rmOCAM, or rmTROP-2.
Mouse
Videos

For Research Use Only