missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKT3 Monoclonal Antibody (A.628.1), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516309
Code : NEW
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with recombinant Akt1 or Akt2.
AKT3 Monoclonal antibody specifically detects AKT3 in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot.Specifications
AKT3 | |
Monoclonal | |
Unconjugated | |
AKT3 | |
AI851531; AKT serine/threonine kinase 3; Akt3; AKT3 kinase; akt3a; D930002M15Rik; DKFZP434N0250; MPPH; MPPH2; Nmf350; PKB gamma; PKBG; PKB-GAMMA; PRKBG; Protein kinase Akt-3; protein kinase B gamma; protein kinase B, gamma; RAC-gamma; RAC-gamma serine/threonine protein kinase; RAC-gamma serine/threonine-protein kinase; RAC-PK-gamma; si:ch211-221n23.2; STK-2; thymoma viral proto-oncogene 3; v-akt murine thymoma viral oncogene 3; v-akt murine thymoma viral oncogene homolog 3; v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma); v-akt murine thymoma viral oncogene homolog 3a; v-akt murine thymoma viral oncogene-like 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10000, 23797, 29414 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
A.628.1 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
Q63484, Q9WUA6, Q9Y243 | |
AKT3 | |
Synthetic peptide corresponding to residues of human Akt3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |