Learn More
AHR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532664
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%); Gorilla, Gibbon, Monkey, Marmoset (95%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
AHR Polyclonal antibody specifically detects AHR in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
AHR | |
Polyclonal | |
Unconjugated | |
AHR | |
Ah; Ah receptor; Ahh; Ahr; Ahre; AH-receptor; aromatic hydrocarbon receptor; aryl hydrocarbon receptor; aryl-hydrocarbon receptor; bHLHe76; Class E basic helix-loop-helix protein 76; dioxin receptor; In | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P35869 | |
AHR | |
Synthetic 20 amino acid peptide from internal region of human Aryl Hydrocarbon Receptor. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |