Learn More
AGTR-2 Mouse anti-Human, Clone: 364805, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3659
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450196
  • £311.64 / 100µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
AGTR-2 Monoclonal antibody MAB3659 Specifically detects AGTR-2 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
AGTR-2
364805
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
AGTR2
HEK293 human embryonic kidney cell line transfected with human AGTR-2 Met1-Ser363 Accession # P50052
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
186
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P50052
AGTR2, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2, AT2ATGR2, AT2R, MRX88, MRX88type-2 angiotensin II receptor
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human AGTR-2. Stains human AGTR-2-transfected cells but not irrelevant transfectants.