Learn More
Agrin Mouse anti-Rat, Clone: 76805, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB550
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460976
  • £337.93 / 500µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Agrin Monoclonal antibody MAB550 Specifically detects Agrin in Rat samples. It is validated for Western Blotting
Specifications
Specifications
Agrin
76805
Western Blot 1 ug/mL
P25304
agrin, agrin proteoglycan, AGRN
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat Agrin in direct ELISAs and Western blots.
Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
AGRN
S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat Agrin Ala1153-Pro1959 (Pro1788-Ser1798 del) Accession # P25304
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
375790
Reconstitute at 0.5 mg/mL in sterile PBS.