missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534749
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (91%), Rat (92%), Chicken (85%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AGR3 Polyclonal antibody specifically detects AGR3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
AGR3 | |
Polyclonal | |
Unconjugated | |
AGR3 | |
AG3; AG-3; AGR3; anterior gradient 3; anterior gradient 3 homolog; anterior gradient 3, protein disulphide isomerase family member; anterior gradient homolog 3; anterior gradient protein 3; anterior gradient protein 3 homolog; BCMP11; Breast cancer membrane protein 11; E030025L21Rik; Gm888; H PDIA18; HAG3; hAG-3; PDIA18; Protein disulfide isomerase family A, member 18; RGD1565406; UNQ642/PRO1272 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
155465 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8TD06 | |
AGR3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 166 of AGR3 (Uniprot ID#Q8TD06) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |