missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-2 Adrenergic Receptor Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518423
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology.
beta-2 Adrenergic Receptor Polyclonal antibody specifically detects beta-2 Adrenergic Receptor in Human samples. It is validated for Western Blot.Specifications
beta-2 Adrenergic Receptor | |
Polyclonal | |
Unconjugated | |
Adrb2 | |
Adrb2; Adrb-2; ADRB2R; ADRBR; Adrenergic beta 2- receptor surface; adrenergic receptor beta 2; adrenergic receptor, beta 2; adrenergic, beta-2, receptor, surface; adrenergic, beta-2-, receptor, surface; adrenoceptor beta 2; adrenoceptor beta 2 surface; adrenoceptor beta 2, surface; AR beta2; B2AR; Badm; BAR; beta 2-AR; beta-2 adrenergic receptor; Beta-2 adrenoceptor; beta-2 adrenoreceptor; beta-2-adrenergic receptor; beta2-adrenergic receptor; beta2-Adrenoreceptor; BETA2AR; catecholamine receptor; Gpcr7; HGNC:286 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
154 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P07550 | |
Adrb2 | |
Synthetic peptide sequence (HQGTVPSDNIDSQ) corresponding to the C-terminus amino acids of ADRB2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |