Learn More
alpha-1b Adrenergic Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532660
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Monkey, Marmoset (95%); Mouse, Hamster, Elephant (85%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
alpha-1b Adrenergic Receptor Polyclonal antibody specifically detects alpha-1b Adrenergic Receptor in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
alpha-1b Adrenergic Receptor | |
Polyclonal | |
Unconjugated | |
Adra1b | |
[a]1b; ADA1B; ADRA1; ADRA1B; Adrenergic alpha 1B- receptor; adrenergic receptor, alpha 1b; adrenergic, alpha-1B-, receptor; adrenoceptor alpha 1B; alpha 1B-adrenoceptor; alpha 1B-adrenoreceptor; alpha-1B adrenergic receptor; Alpha-1B adrenoceptor; alpha-1B adrenoreceptor; alpha-1B-adrenergic receptor; alpha1B-adrenergic receptor; ALPHA1BAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P35368 | |
Adra1b | |
Synthetic 20 amino acid peptide from C-terminal cytoplasmic domain of human ADRA1B. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |