missing translation for 'onlineSavingsMsg'
Learn More

Adiponectin Receptor 2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA11071

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11565912

  • £296.00 / 100µg


EU Stock 1
Add to basket

Description

Description

PA1-1071 detects Adiponectin Receptor 2 from mouse and rat samples.PA1-1071 has been successfully used in Western blot applications. By Western blot, this antibody detects a single ∽43kDa band representing Adiponectin Receptor 2 from mouse liver.The PA1-1071 immunogen is a synthetic peptide corresponding to residues D M F A T P Q Y R G V R of mouse Adiponectin Receptor 2.
Specifications

Specifications

Adiponectin Receptor 2
Polyclonal
Unconjugated
Adipor2
Purified
ADIPOR2
Synthetic peptide corresponding to residues D M F A T P Q Y R G V R of mouse Adiponectin Receptor 2.
Antigen affinity chromatography
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
68465
Western Blot
1mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q8BQS5
ADIPOR2, ACDCR2, Adiponectin receptor protein 2, FLJ21432, MGC4640, PAQR2, Progestin and adipoQ receptor family member II
Rabbit
IgG
100μg
Antibody
Polyclonal
MurineþRat
Videos